Masterflex® Vortex Flowmeters with Temperature for Liquids, Avantor®
Leverandør: Avantor Fluid Handling
32825-68 32825-70 32825-72 32825-74
MFLX32825-76EA 4400 NOK
MFLX32825-76 MFLX32825-68 MFLX32825-70 MFLX32825-72 MFLX32825-74
Masterflex® Vortex Flowmeters with Temperature for Liquids, Avantor®
Flowmetere
Built-in 1000 Ω RTD sensor provides direct temperature measurement of media.
  • Deigned with rugged PPA molded construction for enhanced durability
  • Excellent media compatibility with ETFE, EPDM, and PPD wetted parts
  • Use with low viscosity liquids for laboratory and industrial applications
These economical vortex flowmeters are suitable for water and water glycol-based heat exchange systems. The transmitters work on the vortex shedding flow principle. As flow velocity increases, alternating vortices are formed on each side of the bluff body within theMore Product Information