Burkert Paddle-Wheel Sensors with Integral Fittings
Leverandør: Avantor Fluid Handling
33117-15
MFLX33117-15EA 6250 NOK
MFLX33117-15
Burkert Paddle-Wheel Sensors with Integral Fittings
Flytindikatorer
Enhance accuracy of your flow readings.
  • Can be used with various liquids
These paddle-wheel sensors are ideal for measuring both neutral and aggressive, solid-free liquids. Built-in pipe fittings ensure simple installation into pipes from 1/2" up to 2". Mount sensor/fittings horizontally or vertically.
Choose from polyvinyl chloride (PVC), brass, or stainless steel (SS) sensor/fittings. PVC models come with true union fittings (solvent/fusion connection). Order brass and SS models with NPT(F) fittings for high temperature or high pressure applications.More Product Information