John Guest Polypropylene Push-To-Connect Male Threaded Adapter Fittings
Leverandør: Avantor Fluid Handling
34007-56
MFLX34007-53EA 185 NOK
MFLX34007-53 MFLX34007-54 MFLX34007-55 MFLX34007-56
John Guest Polypropylene Push-To-Connect Male Threaded Adapter Fittings
Fittings og kontakter
Quick, leakproof connection.
  • Polypropylene and EPDM materials meet NSF 51, and NSF 61 requirements for food-grade applications, and is FDA compliant
Push-to-connect fittings offer secure connections and quick disconnection. Simply push tubing past the O-ring to the internal tube stop—steel teeth on the collet grip tubing for leakproof connections. To disconnect, press collet against fitting body to disengage steel teeth—tubing slides right out.
Fittings work with semi-rigid plastic (polypropylene, polyethylene, nylon) and soft metal (brass, copper,More Product Information