Masterflex® 3/4" Sanitary Clamp Low Flow Male Quick-Disconnect Fittings, Avantor®
Leverandør: Avantor Fluid Handling
MFLX40257-40EA 1000 NOK
MFLX40257-40 MFLX40257-41 MFLX40257-47 MFLX40257-48 MFLX40257-54 MFLX40257-55 MFLX40257-61 MFLX40257-62 MFLX40257-68 MFLX40257-69 MFLX40257-75 MFLX40257-76
Masterflex® 3/4" Sanitary Clamp Low Flow Male Quick-Disconnect Fittings, Avantor®
Fittings og kontakter
Mechanical innovations reduce risk of fluid cross-over and contamination. A mating connection is made with a Low Flow Female fitting.
  • Aseptic connect and disconnect
  • Spring-less fluid path
  • Recessed mating features
  • USP Class VI Polypropylene and PVDF housing materials
  • Elimination of accidental disconnect with safety slide release
Masterflex® quick-disconnect fittings are an innovative and aseptic connection that allows a larger volume of fluid to pass through at lower line pressures in a smaller body. As the overall quick disconnect design has matured,More Product Information