Masterflex® Adapter Fittings, Luer to Luer, Nylon, Avantor®
Leverandør: Avantor Fluid Handling
45502-22 45502-28 45505-56
MFLX45502-56EA 370 NOK
MFLX45502-56 MFLX45502-80 MFLX45502-84 MFLX45502-85 MFLX45505-72 MFLX45502-22 MFLX45502-28 MFLX45505-56
Masterflex® Adapter Fittings, Luer to Luer, Nylon, Avantor®
Fittings og kontakter
Easily transition to a luer fitting.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs,