316 SS Heavy-Duty Sanitary Centrifugal Pumps
Leverandør: Avantor Fluid Handling
61133-54
MFLX61133-54EA 76650 NOK
MFLX61133-54
316 SS Heavy-Duty Sanitary Centrifugal Pumps
Pumper Sentrifugalpumper
Clean-in-place (CIP) 3A sanitary transfer pump.
  • 316L stainless steel liquid end construction
  • Ideal for RO water and pharmaceutical manufacturing
  • Casing is twice as thick as the 76710 series of pumps
  • Designed for steam-in-place (SIP)
Piping changes are eliminated due to the interchangeable design that these economical pumps offer when compared to other pumps. Models are available with inlets up to 6" contact our Application Specialists for assistance in sizing the proper pump for your system.
Leveringsinformasjon: Includes conduit box for hardwiring.