High-Efficiency Polypropylene Standard Centrifugal Pumps
Leverandør: Avantor Fluid Handling
72009-30
MFLX72009-30EA 35100 NOK
MFLX72009-30
High-Efficiency Polypropylene Standard Centrifugal Pumps
Pumper Sentrifugalpumper
Split volute casing forms a volute chamber to increase pump efficiency, reduce horsepower, and energy usage.
  • Pumps can run dry for up to one hour
  • To reduce maintenance, bearings are kept cool with heat dissipation vent holes and a large flush groove
  • Use in production processes such as filtering, spraying, washing, and etching
Pumps are set up for hard-wiring; cord and plug are not included.
Leveringsinformasjon: Includes conduit box for hardwiring.