Continuous-Duty Double-Diaphragm Pumps
Leverandør: Avantor Fluid Handling
75102-00
MFLX75102-00EA 0 NOK
MFLX75102-00
Continuous-Duty Double-Diaphragm Pumps
Pumper Membranmålepumper
No metal wetted parts permit use with corrosive liquids.
  • Continuous at 15 psi (1 bar) and 70 °F (21 °C) ambient temperature
  • All models feature factory-set pressure limits to protect pumps from damage—pumping stops when high limit is reached, resumes when pressure decreases to low setting.
Multi-chambered pump offers efficient transfer of mild acids, bases, and light oils. Thermal overload protected, permanent-magnet motor. Pump includes 6-ft power cord with three-prong plug. Order replacement diaphragms or valve kits under accessories.