Flexible Impeller Self-Priming Non-Metallic Pumps
Leverandør: Avantor Fluid Handling
75421-00 75501-12
MFLX75421-00EA 0 NOK
MFLX75421-00 MFLX75501-12
Flexible Impeller Self-Priming Non-Metallic Pumps
Pumper Flexible/Rotary/Roller Impeller Pumps
Pumps fluids in applications where durability is needed—the 12 VDC models are ideal for remote/portable use.
  • Pharmaceutical—processing lab solutions, lotions, preparations, and filtering solutions
  • Industrial—circulating and transferring corrosive liquids, transferring solutions with entrained air (such as soaps and detergents), sampling, and pilot plant installations
  • Agricultural—transferring liquid fertilizers, herbicides, and pesticides
  • Metal plating—filtration of plating solutions, filling, and emptying vats
These flexible impeller self-priming non-More Product Information