Masterflex® Two-Stage High-Pressure Centrifugal Pumps, Avantor®
Leverandør: Avantor Fluid Handling
75506-70
MFLX75506-70EA 33975 NOK
MFLX75506-70
Masterflex® Two-Stage High-Pressure Centrifugal Pumps, Avantor®
Pumper Sentrifugalpumper
Two-stage design gives superior output pressures.
  • Close-coupled design saves space; simplifies maintenance and installation
  • Fabricated from 304 stainless steel for corrosion resistance
  • Back pullout design allows servicing without disturbing piping.
Pumps are non-overloading and rated for continuous duty throughout entire operating range for reliable performance and long life. Install either vertically or horizontally.