White Epoxy Motor Sanitary Centrifugal Pumps
Leverandør: Avantor Fluid Handling
76710-10
MFLX76710-10EA 38100 NOK
MFLX76710-10
White Epoxy Motor Sanitary Centrifugal Pumps
Pumper Sentrifugalpumper
Clean-in-place (CIP) 3A sanitary transfer pump.
  • Washdown motors available in white epoxy
  • Stainless steel adapters are standard
  • Ideal for RO water and pharmaceutical manufacturing
Piping changes are eliminated due to the interchangeable design that these economical pumps offer when compared to other pumps. Models are available with inlets up to 6" contact our Application Specialists for assistance in sizing the proper pump for your system.
Leveringsinformasjon: Includes conduit box for hardwiring.