Masterflex® B/T® Rapid-Load Pump Heads, Avantor®
Leverandør: Avantor Fluid Handling
MFLX77111-50EA 70175 NOK
MFLX77111-50 MFLX77111-55
Masterflex® B/T® Rapid-Load Pump Heads, Avantor®
Pumper Pumpehoder Peristaltiske pumpehoder
Convert your existing motor into a high-capacity Masterflex® B/T® drive.
  • Newly designed occlusion mechanism slides occlusion bed out of the way for easy tube loading and improved tubing occlusion
  • Accepts B/T PerfectPosition™ pump tubing sizes B/T 87 and B/T 91
  • Interlock switch shuts off power to stop drive when door is opened
  • Three rollers and 5.6:1 hear ratio provides optimal flow capacity
  • Nylatron® rotor / roller for greater durability and chemical resistance
Enjoy all the advantages of peristaltic tubing drives using your existing motor. GentleMore Product Information