Masterflex® I/P® Easy-Load® Pump Heads for Precision Tubing, Avantor®
Leverandør: Avantor Fluid Handling
MFLX77601-60EA 15725 NOK
MFLX77601-60 MFLX77602-00 MFLX77602-10 MFLX77602-30 MFLX77602-80
Masterflex® I/P® Easy-Load® Pump Heads for Precision Tubing, Avantor®
Pumper Pumpehoder Peristaltiske pumpehoder
Minimize downtime with faster tubing changes.
  • New design and automatic tubing retention allow for multiple tubing sizes—change tubing without removing head from drive
  • Three roller design for smooth flow
  • Choose from stainless steel (SS) or cold-rolled steel (CRS) rotor / roller for greater durability and chemical resistance
  • Pump heads are stackable for multichannel applications
  • Compact design for efficient use of space
  • Accept multiple tubing sizes for maximum flexibility
  • Automatic tubing retention eliminates user adjustments and makes setup easy—offers