Masterflex® Ismatec® Minicartridge Pump Heads for 3-Stop Tubing, Avantor®
Leverandør: Avantor Fluid Handling
MFLX78002-90EA 16375 NOK
MFLX78002-90
Masterflex® Ismatec® Minicartridge Pump Heads for 3-Stop Tubing, Avantor®
Pumper Pumpehoder Peristaltiske pumpehoder
Easily connect to your Ismatec® drive.
  • Snap your pump head onto your drives quickly with turn-and-lock mounting system
  • Select from sixteen interchangeable pump heads that offer flow rates from 0.01 to 3660 ml/min
  • RS-232 interface connects to your PC
Designed specifically to be used with Ismatec 3-stop tubing.