Masterflex® Ismatec® Reglo Digital Piston Pump Drive, Avantor®
Leverandør: Avantor Fluid Handling
MFLX78018-60-EUEA 49250 NOK
MFLX78018-60-EU
Masterflex® Ismatec® Reglo Digital Piston Pump Drive, Avantor®
Pumper Pumpedrev Stempelpumpedrift
Achieve high pressure, accuracy, and repeatability -- with an intuitive touch-screen interface.
  • Compact, benchtop pumps optimize ease of use, accuracy, and reliability
  • Compatible with FMI RH series piston pump heads for flow ranges from 0.025 to 180 ml/min
  • 5” (12.7 cm) capacitive TFT/RGB touch screen for crisp resolution and a wide viewing angle
  • Flexible operational control via screen or analog remote control
  • Continuous run, volume dispense, and time dispense modes -- batch control and interval/off time in Volume and Time modes
  • Pump lets you save and