Masterflex® Ismatec® Pump Tubing, 3-Stop Color-Coded Autoanalysis/Microbore, Platinum-Cured Silicone, Avantor®
Leverandør: Avantor Fluid Handling
95603-24 95603-38
MFLX95603-24EA 1625 NOK
MFLX95603-24 MFLX95603-26 MFLX95603-28 MFLX95603-38 MFLX95603-44 MFLX95603-48 MFLX95603-40 MFLX95603-42 MFLX95603-22 MFLX95603-30 MFLX95603-34 MFLX95603-36 MFLX95603-32
Masterflex® Ismatec® Pump Tubing, 3-Stop Color-Coded Autoanalysis/Microbore, Platinum-Cured Silicone, Avantor®
Slanger Peristaltisk pumpeslange
Designed for optimal operation in your Ismatec pumps.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • No leachable additives or plasticizers
To complete your Ismatec® pumping system, order three-stop color-coded tubing and extension tubing. Tubing comes in various formulations for almost anyMore Product Information