Masterflex® Ismatec® Pump Tubing, GORE® STA-PURE® Series PFL Single Elements, Avantor®
Leverandør: Avantor Fluid Handling
MFLX96252-00EA 11825 NOK
MFLX96252-00 MFLX96252-02 MFLX96252-06 MFLX96252-10 MFLX96252-12 MFLX96252-16 MFLX96252-18 MFLX96252-24 MFLX96252-26 MFLX96252-30 MFLX96252-34 MFLX96252-40 MFLX96252-04 MFLX96252-08 MFLX96252-22 MFLX96252-28 MFLX96252-38
Masterflex® Ismatec® Pump Tubing, GORE® STA-PURE® Series PFL Single Elements, Avantor®
Slanger Peristaltisk pumpeslange
Excellent choice for high pressure, low spallation peristaltic tubing applications.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Engineered for long life in peristaltic pump applications
  • Unique composite of expanded PTFE and platinum-cured perfluoroelastomer
Applications: Chemical, criticalMore Product Information