Masterflex® Ismatec® Pump Tubing, 2-Stop Microbore, Chem-Durance® Bio, Avantor®
Leverandør: Avantor Fluid Handling
MFLX96327-14EA 1725 NOK
MFLX96327-14 MFLX96327-26 MFLX96327-30 MFLX96327-34 MFLX96327-36 MFLX96327-42 MFLX96327-48 MFLX96327-18
Masterflex® Ismatec® Pump Tubing, 2-Stop Microbore, Chem-Durance® Bio, Avantor®
Slanger Peristaltisk pumpeslange
Designed for optimal operation in your Ismatec pumps.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Long service life - Outstanding fatigue resistance due to flexing!
  • Suitable for applications using acetone, methyl-ethyl-ketone (MEK), and other highly aggressive chemicals
  • Appearance: Opaque