Masterflex® L/S® Spooled Pump Tubing, Peroxide-Cured Silicone, Avantor®
Leverandør: Avantor Fluid Handling
MFLX96407-13EA 7150 NOK
MFLX96407-13 MFLX96407-14 MFLX96407-15 MFLX96407-16 MFLX96407-17 MFLX96407-18 MFLX96407-24 MFLX96407-25
Masterflex® L/S® Spooled Pump Tubing, Peroxide-Cured Silicone, Avantor®
Slanger Peristaltisk pumpeslange
Excellent biocompatibility for a wide range of applications.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and