Masterflex® L/S® High-Performance Precision Pump Tubing, BioPharm Platinum-Cured Silicone, Avantor®
Leverandør: Avantor Fluid Handling
MFLX96420-15EA 3575 NOK
MFLX96420-15 MFLX96420-24 MFLX96420-35 MFLX96420-36
Masterflex® L/S® High-Performance Precision Pump Tubing, BioPharm Platinum-Cured Silicone, Avantor®
Slanger Peristaltisk pumpeslange
Ultra-smooth inner surface minimizes particle entrapment.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for