Masterflex® I/P® Spooled Pump Tubing, BioPharm Platinum-Cured Silicone, Avantor®
Leverandør: Avantor Fluid Handling
MFLX96423-26EA 70750 NOK
MFLX96423-26 MFLX96423-73 MFLX96423-88 MFLX96423-89
Masterflex® I/P® Spooled Pump Tubing, BioPharm Platinum-Cured Silicone, Avantor®
Slanger Peristaltisk pumpeslange
Smooth inner surface designed to reduce risk of particle entrapment and contamination.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory Approvals to match your unique application
Choose our BioPharm SiliconeMore Product Information