Masterflex® Stainless Steel Fittings, Avantor®
Leverandør: Avantor Fluid Handling
MFLX96881-20EA 275 NOK
MFLX96881-20
Masterflex® Stainless Steel Fittings, Avantor®
Klemmer Slangeklemmer
Assure the effectiveness of your steam-in-place (SIP) processes.
  • Easily incorporate into existing tubing or piping lines
  • System uses standard sanitary Tri-Clamp® fittings
  • Results can be read in only one hour
The Attest in-line sterilty testing system allows you to determine the effectiveness of SIP efforts. A small ampule containing steam-sensitive bacteria is inserted into a holder gasket. The gasket can then be mounted into standard sanitary Tri-Clamp connections and capped with an end cap. After a SIP or CIP procedure, the ampule is analyzed in oneMore Product Information