Masterflex® I/P® High-Performance Precision Pump Tubings, Puri-Sil™ Platinum-Cured Silicone
Leverandør: Avantor Fluid Handling
MFLX97411-88EA 14950 NOK
MFLX97411-88
Masterflex® I/P® High-Performance Precision Pump Tubings, Puri-Sil™ Platinum-Cured Silicone
Slanger Peristaltisk pumpeslange
Up to five times longer life and lower spallation provides enhanced consistency over time.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory Approvals to match your unique application
Choose our Puri-Sil™More Product Information