Masterflex® High-Purity Plastic Diaphragm Valves, Avantor®
Leverandør: Avantor Fluid Handling
98520-00
MFLX98520-00EA 9700 NOK
MFLX98520-00
Masterflex® High-Purity Plastic Diaphragm Valves, Avantor®
Ventiler Membranventiler
Fully drainable and easy to maintain!
  • Eliminate leaks—ideal for food or pharmaceutical processing
  • Compatible with Tri-Clamp® connections for easy maintenance, change out, and orientation—no cutting or welding needed!
  • Choose either natural polypropylene (PP) or Kynar® (PVDF) body—both are excellent for ultrapure applications
  • Models with Kynar® (PVDF) body offer additional chemical resistance to corrosives
These high-purity valves feature manual actuators. Maximum viscosity is 400 cSt. Maximum fluid temperature is 180 °F (82 °C).