Bucket for use with wringer
Leverandør: MICRONOVA
B-3
MICRB-3EA 33950 NOK
MICRB-3
Bucket for use with wringer
Bøtter
Durable stainless steel bucket is compatible with W-6 wringer for an all electropolished stainless steel system.
  • With casters (2")
  • With handle
  • Round
Bestillingsinformasjon: Wringer has to be ordered separately.