UV-Vis wavelenth standard
Leverandør: MICROSOLV
PQA-HOX-01
MICSPQA-HOX-01EA 3375 NOK
MICSPQA-HOX-01
UV-Vis wavelenth standard
Standarder Fargereferanse og spekroskopistandarer
Wavelength calibration standard solution for UV-Vis detectors is composed of 4% Holmium oxide in 10% Perchloric acid.