Amicon™ XM Ultrafiltration Membranes Dics Filters
Leverandør: Merck
14112AM
MILF14112AMEA 3525 NOK
MILF14112AM
Amicon™ XM Ultrafiltration Membranes Dics Filters
Filter Membranfilter
Ultrafiltration membranes - Amicon™ XM polyacrylonitrile/PVC co-polymer filters. High-recovery membranes for stirred cells.
  • Regenerated cellulose membranes
  • For concentration of hydrophobic samples or to desalt extremely dilute solutions
Hydrophilic, tight microstructure assures high retention with low adsorption of protein, DNA, or other macromolecules.