Upstate® Autorad Orientation Markers for Western Blotting
Leverandør: Merck
20-135
MILF20-135EA 1600 NOK
MILF20-135
Upstate® Autorad Orientation Markers for Western Blotting
Elektroforese reagenser Molekylvekt markører
Autorad orientation markers for use on immunoblots in detection of Chemiluminescence in Western blotting, dot/slot blotting.