Chemicon® ReBlot Plus Strong Antibody Stripping Solution, 10X
Leverandør: Merck
2504
MILF2504EA 2650 NOK
MILF2504
Chemicon® ReBlot Plus Strong Antibody Stripping Solution, 10X
Elektroforese reagenser Western Blotting Reagents
Western blotting is a commonly used technique for studying protein function and localisation. Typically, protein samples are electro-phoresed on SDS-page and transferred to a membrane such as nitrocellulose or nylon, where they are probed with specific antibodies. Unlike nucleic acid based technologies, which allow reuse of southern and northern blots, it has been difficult to reuse western blots.
  • Detection method: Chemiluminescence
  • Strong antibody stripping solution (10X) - (2 containers, 25 ml each)
The MILLIPORE re-blot plus western blot strong antibody stripping solution is effective for removal of antibodies from western blots that have been developed with chemiluminescence or radioactive iodine or other isotopes. It is not recommended for stripping colorimetric substrates (TMB, DAB, 4-chloronapthol, and more), as it is not possible to effectively remove substrates that precipitate at the reaction site.
The re-blot plus western blot strong antibody stripping solution should be used only for qualitative purposes until it has been established by comparative blot analysis that stripping does not quantitatively affect a given antigen.
NB: This product is for research use only not for diagnostic or in vivo use.