Chemicon® BacterialXpress™ Nucleic Acid Extraction Kit

Leverandør: Merck

3096
MILF3096EA 0 NOK
MILF3096
Chemicon® BacterialXpress™ Nucleic Acid Extraction Kit
Nukleinsyre reagenser Nukleinsyrerensingsreagenser og -kit

BacterialXpress™ Nucleic acid extraction kit is intended for the purification of bacterial DNA from plasma, serum, cerebral spinal fluid, amniotic fluid, tissue, bone marrow, and cell culture.


The BacterialXpress™ kit provides a rapid method for the extraction and purification of amplification-competent nucleic acids; however, the kit does not separate DNA from RNA. The BacterialXpress™ nucleic acid extraction kit can be used for the extraction and purification of a broad range of bacterial DNA from a variety of sources. However, performance cannot be guaranteed for all bacteria from all sources.


Suitable for the purification of bacterial DNA from plasma, serum, cerebral spinal fluid, amniotic fluid, tissue, bone marrow, and cell culture.


Leveringsinformasjon: Supplied woth Bacterial DNA extraction reagent (11 ml).

Bestill nå

SPESIFIKASJONER

Environmentally Preferable
Applikasjon
Target molecule
Refine target molecule
Sample type
Method / Format

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR