Quartz fiber filters, type AQFA
Leverandør: Merck
AQFA03700 AQFA04700 AQFA09050 AQFA11050 AQFA8X105
MILFAQFA03700EA 2400 NOK
MILFAQFA03700 MILFAQFA04700 MILFAQFA09050 MILFAQFA11050 MILFAQFA8X105
Quartz fiber filters, type AQFA
Filter Glassfiberfilter
Made of very pure quartz fibers with no glass fibers or binders. This makes these filters well suited for measuring heavy metal concentrations and small amounts of particles (such as the USEPA PM 10 ambient air monitoring method). The filters also exhibit good weight and form stability.