EZ-Fit™ Manifold
Leverandør: Merck
EZFITMIC03
MILFEZFITMIC03EA 46375 NOK
MILFEZFITMIC03
EZ-Fit™ Manifold
Manifold
EZ-Fit™ Manifold for universal laboratory filtration.
  • Unique design for easy cleaning and decontamination
  • Different filtration heads for both reusable and disposable filtration devices
  • Quick-fit connectors for the vacuum tubing
  • Low height for ease of use in laminar flow hoods
  • Each component can be removed quickly and easily without any additional tools
  • A check valve is integrated into the tubing connection to prevent liquid back-flow from the vacuum system
  • The 2-way valve prevents the user from accidentally touching the filtration head while opening or