Millipore® Immobilon®-NC Hydrophilic Transfer Membrane
Leverandør: Merck
HATF85R
MILFHATF85REA 4325 NOK
MILFHATF85R
Millipore® Immobilon®-NC Hydrophilic Transfer Membrane
Overføringsmembraner
Triton-free, mixed cellulose esters transfer membrane.
  • Western blotting
  • Electrostatic
  • Hydrophilic
  • Mixed cellulose esters (MCE)
  • Plain/white filter
  • 1 roll, 8,5 cm × 10 m
  • 0,45 µm pore size
Detection method: chromogenic, radioactive and fluorescent.
Compatible stains: Amido black, India ink, Ponceau-S red, Colloidal gold and CPTS.