Multiscreen® 96 Well Plate, PTFE Membrane
Leverandør: Merck
MDRPN0410
MILFMDRPN0410EA 11300 NOK
MILFMDRPN0410
Multiscreen® 96 Well Plate, PTFE Membrane
Mikroplater Deepwell mikroplater
The MultiScreen® deep well solvinert filter plate is intended for a wide range of applications including total drug analysis, bead cleavage (solid-phase combinatorial chemistry and peptide synthesis), and sample preparation before high-performance liquid chromatography (HPLC)/ultraviolet (UV) and liquid chromatography-tandem mass spectrometry (LC-MS/MS). This filter plate is fully automation compatible.
  • H: 40,7 mm
  • L: 128 mm
  • W: 85,5 mm
  • Filtration area: 0,28 cm²
  • Hydrophobic
  • Ambient
MultiScreen® deep, 96 well solvinert has been used to produce adsorptionMore Product Information