MultiScreen® HTS Filter Plate, Plasmid

Leverandør: Merck
MSNUPSD50
MILFMSNUPSD50EA 0 NOK
MILFMSNUPSD50
MultiScreen® HTS Filter Plate, Plasmid
Mikroplater Filterplater
Multiscreen HTS filter plate, plasmid also called as Montage Plasmid Miniprep.

  • Automation compatible
  • Suitable for DNA purification
  • Shipped in: Ambient
  • Quality level: 400

Application: Research category and nucleic acid
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR