Millipore® Immobilon®-FL NOW Bundle Hydrophobic PVDF Transfer Membrane
Leverandør: Merck
NWKITFL
MILFNWKITFLEA 13925 NOK
MILFNWKITFL
Millipore® Immobilon®-FL NOW Bundle Hydrophobic PVDF Transfer Membrane
Overføringsmembraner
Hydrophobic PVDF transfer membrane for western blotting using fluorescent, chemifluorescence, chromogenic, or chemiluminescence.
  • Western blotting
  • Hydrophobic
  • Plain/white filter
  • 0,45 µm pore size
  • Comes in a 8,5 cm × 10 m roll
The Immobilon NOW system offers 1-cut convenience for standard mini-blots (7 × 8,5cm) and midi-blots (8,5 × 13,5cm).
Compatible stains: All fluorescence dyes, Coomassie brilliant blue, Amido black, Ponceau-S red and Sypro® ruby