Millipore® Immobilon®-E NOW Bundle Hydrophobic PVDF Transfer Membrane
Leverandør: Merck
NWKITIE
MILFNWKITIEEA 13925 NOK
MILFNWKITIE
Millipore® Immobilon®-E NOW Bundle Hydrophobic PVDF Transfer Membrane
Overføringsmembraner
The first PVDF transfer membrane for western blotting that wets out in transfer buffer or water. For chemiluminescence or chromogenic detection.
  • Western Blotting
  • Immunoblotting (Western)
  • Dot Blot
  • White/plain filter
  • Electrostatic, hydrophobic
  • Easily wetted with water
Immobilon-E is the first PVDF membrane for Western blotting that can be wetted with with aqueous transfer buffer. This eliminates the alcohol pre-wet step required with other PVDF membranes.
45 µm pore size recommended for most Western blotting applications, especially proteins greater