Millipore® Immobilon®-NC NOW Bundle Hydrophilic Mixed Cellulose Esters Transfer Membrane
Leverandør: Merck
NWKITNC
MILFNWKITNCEA 16350 NOK
MILFNWKITNC
Millipore® Immobilon®-NC NOW Bundle Hydrophilic Mixed Cellulose Esters Transfer Membrane
Overføringsmembraner
Triton-free, mixed cellulose esters transfer membrane, includes 1 dispenser.
  • Radioactive, chromogenic and fluorescent detection method
  • Western blotting
  • Plain/white filter
  • Hydrophilic
  • 2 rolls, 8,5 cm × 10 m
  • 0,45 µm pore size
This bundle includes the Immobilon NOW dispenser and two 8,5 cm × 10 m rolls of Immobilon-NC transfer membrane. The Immobilon NOW system offers 1-cut convenience for standard mini-blots (7×8,5 cm) and midi-blots (8,5×13,5 cm).
Compatible stains: Amido black, CPTS, Colloidal gold, India ink and Ponceau-S red.