Millicell® EZ Slide Holder
Leverandør: Merck
PEZXMSH01
MILFPEZXMSH01EA 2700 NOK
MILFPEZXMSH01
Millicell® EZ Slide Holder
Objektglassholdere
Microscope slide holder for 4 and 8 well units, holds two slides.
  • Shipped in ambient temperature
Techniques: cell based assay: suitable cell culture; mammalian: suitable immunocytochemistry: suitable