Millicell® High Yield Multilayer Culture Flasks
Leverandør: Merck
PFHYS1008 PFHYS0616
MILFPFHYS1008EA 10325 NOK
MILFPFHYS1008 MILFPFHYS0616
Millicell® High Yield Multilayer Culture Flasks
Kolber Celledyrkningsflasker
When growing large numbers of cells, you need a high throughput flask to save space and time. Each Millicell HY (high yield) flask provides a consistent, quality growth environment across all layers, with the same volume of media in each layer. Recovering cells is as easy as growing them.
  • Consistent, high cell yields and viability
  • Same media requirements (volume per unit area) as traditional T-flasks
  • Uniform culture conditions without media leaking or spillover between layers
With easy access to pipetting or pouring, Millicell HY flasks deliver