Millicell® 96-Well Cell Culture Insert Plates
Leverandør: Merck
PSHT004S5 PSRP004R5 PSHT004R1 PSHT004R5 PSRP004R1
MILFPSHT004S5EA 32075 NOK
MILFPSHT004S5 MILFPSRP004R5 MILFPSHT004R1 MILFPSHT004R5 MILFPSRP004R1
Millicell® 96-Well Cell Culture Insert Plates
Mikroplater Celledyrkningsplater
Millicell cell culture insert plates are a patented device designed to support cell growth and differentiation of endothelial and epithelial cell lines, including CaCo-2.
  • Isopore™ matrix
  • 0,11 cm² filtration area
  • Tissue Culture (TC)-treated surface binding type
  • Dimension: 84,8×128×26,2 mm
Seed-and-feed systems
Automated liquid handling systems and span pipettes
Basolateral and apical access to cells
Transepithelial electrical resistance (TEER) measurement systems
The Millicell device is available with a 0,4 µm PCF and a variety of PET membrane poreMore Product Information