SNAP i.d. 2.0 Protein Detection Systems Mini
Leverandør: Merck
TISNAP2MINI
MILFTISNAP2MINIEA 0 NOK
MILFTISNAP2MINI
SNAP i.d. 2.0 Protein Detection Systems Mini
Analysatorer Proteinanalysatorer
Developed to meet the needs of our Western blotting customers, the SNAP i.d. 2.0 system produces blots of a very high quality. Unique vacuum-driven technology and a built-in flow distributor actively drive reagents through the membrane.
Bestillingsinformasjon: This system includes the SNAP i.d. 2.0™ base, two mini blot holding frames, two antibody collection trays, vacuum tubing, rolling pad and blot roller, two wetting trays and a quick start guide.