Solvent-Resistant Stirred Cell for 76 mm Membranes

Leverandør: Merck
XFUF07601
MILFXFUF07601EA 43925 NOK
MILFXFUF07601
Solvent-Resistant Stirred Cell for 76 mm Membranes
Omrørere Magnetrørere
For applications requiring compatibility with solvents, this Millipore stirred cell is available in two sizes. Certain aldehydes, ketones (e.g., acetone) and aliphatic ethers/esters may reduce the life of the fluorocarbon O-rings.

  • Borosilicate glass cylinder and PTFE components for broad compatibility
  • Autoclavable with membrane in-place
  • Top plate opening provides access to contents without dismantling
  • Few components, so its easy to clean and assemble
  • For 47 mm disc filters

Applications are concentration and buffer exchange involving organic solvents.
More Product Information
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR