Fluid Sampling Kit
Leverandør: Merck
XX6403730
MILFXX6403730EA 53250 NOK
MILFXX6403730
Fluid Sampling Kit
Prøvetakere Væskeprøvetakere
Fluid sampling kit for sampling turbine fuels, water or other liquids through Millipore contamination analysis monitors.
  • Sturdy carrying case and stainless steel sampling assembly provide years of in-field service
  • Fluid Sampling Kit includes 12 Tenite contamination monitors
The Fluid Sampling Kit is a complete solution for field-based fluid collection and contamination analysis. The stainless steel sampler assembly plugs into a quick-release valve installed in the system line, which allows a measured quantity of liquid to pass through the monitor. AMore Product Information