Milli-Q® SQ 200 Water Purification System
Leverandør: Merck
ZSQ200U0T0
MILLZSQ200U0T0EA 39550 NOK
MILLZSQ200U0T0
Milli-Q® SQ 200 Water Purification System
Vannrensesystemer Rene og ultrarene vannsystemer
The Milli-Q® SQ 200 water purification system delivers ultrapure (Type 1) water from an SQ Switch 3,5 L tank filled with pure reverse osmosis (RO) water.
  • Simple, manual handset dispenser allows any new or untrained user to deliver ultrapure water at variable flow rates, from 1,6 L/min to drop by drop
  • Reliable 18,2 MΩ·cm resistivity ultrapure (Type 1) water quality is indicated by a colored LED, and TOC is reduced by an integrated photo-oxidation UV lamp
  • This autonomous unit provides freshly dispensed ultrapure water anywhere with an electrical outlet