Milli-Q® SQ 200P Purification System
Leverandør: Merck
ZSQ200UPT0
MILLZSQ200UPT0EA 77125 NOK
MILLZSQ200UPT0
Milli-Q® SQ 200P Purification System
Vannrensesystemer Rene og ultrarene vannsystemer
The Milli-Q® SQ 200P water purification system delivers ultrapure (Type 1) water from a lab's pressurized purified (RO/DI) water loop. It benefits from a flexible handset dispenser to deliver ultrapure water.
  • Ultrapure (Type 1) water dispensing module from pressurised purified water loop
  • Flexible dispensing
  • For elementary lab use
  • Compact - mobile - scalable
  • Simply plug in, use and maintain
Simple, manual handset dispenser allows any new or untrained user to deliver ultrapure water at variable flow rate, from up to 1,6 L/min to drop by drop. Reliable 18More Product Information