Milli-Q® SQ 240C Purification System
Leverandør: Merck
ZSQ240UCT0
MILLZSQ240UCT0EA 0 NOK
MILLZSQ240UCT0
Milli-Q® SQ 240C Purification System
Vannrensesystemer Rene og ultrarene vannsystemer
The Milli-Q® SQ 240C water purification system delivers both pure reverse osmosis (RO) water and ultrapure (Type 1) water. It benefits from a flexible handset dispenser to deliver ultrapure water. It can also be equipped with a gravity valve (optional) to dispense pure RO water directly from the SQ Switch 3,5 L tank base.
  • Combined pure RO water production station and ultrapure (Type 1) water dispensing module for one 3,5 L switch tank
  • Flexible dispensing
  • For elementary lab use
  • Compact, mobile - scalable
  • Simply plug in, use and maintain
This Milli-Q®More Product Information