Milli-Q® SR 240L Purification System
Leverandør: Merck
ZSR240RLT0
MILLZSR240RLT0EA 58625 NOK
MILLZSR240RLT0
Milli-Q® SR 240L Purification System
Vannrensesystemer Rene og ultrarene vannsystemer
The Milli-Q® SR 240L water purification system delivers pure reverse osmosis (RO) water to two mobile SQ Switch 3,5 L tanks.
  • Gravity Valve (optional) lets you dispense RO water at up to 6 L/h directly from the SQ Switch tank base
  • Compact setup and self-guided installation manual and videos let you plug the system wherever is convenient for your lab activity and move it later if needed
  • Individual Twist and Lock cartridge replacement simplifies maintenance and limits running cost
  • Compact size plus water and energy savings support lab sustainability efforts