Milligard® Membrane Filter Units, PES 1,2/0,8 μm, Opticap® T-Line 30 Capsule
Leverandør: Merck
KMP8G3TTT1
MILXKMP8G3TTT1EA 24825 NOK
MILXKMP8G3TTT1
Milligard® Membrane Filter Units, PES 1,2/0,8 μm, Opticap® T-Line 30 Capsule
Filter Membranfiltertrakter, sett
Milligard® PES 1.2/0.8 μm filters are designed for reliable particulate removal from a broad range of fluid streams. When used upstream of sterilizing-grade filters or other unit operations, they improve processing efficiency. Milligard® PES filters are supported by the Emprove® Program which consolidates product specific testing, quality and regulatory information to simplify compliance needs.
  • Mode of Action: Filtration (size exclusion)
  • Application: BioProcessing
  • Intended Use: Reduction or removal of microorganism/bioburden
  • Instructions for