Robotspiss for Tecan®
Leverandør: Thermo Fisher Scientific
901-262 904-251 901-252 903-262 903-261 903-021 904-262 904-261 904-021 901-261 902-262 902-261 902-021 902-253 903-011G 903-251G 903-252G 904-011G 904-251G 904-252G 903-252 903-251 903-011 904-252 904-011 901-251 901-011 902-252 902-251 902-011 901-253 903-253 904-253 901-021 908-261 907-261 908-021 908-262 907-262 902-262XP 906-262 906-261 902-261XP 906-021 907-021 903-033
736-0045EA 2625 NOK
736-0045 736-0061 736-0043 736-0058 736-0010 736-0009 736-0066 736-0012 736-0011 736-0006 736-0050 736-0008 736-0007 736-0049 736-0052 736-0054 736-0056 736-0060 736-0062 736-0064 736-0055 736-0053 736-0051 736-0063 736-0059 736-0042 736-0041 736-0048 736-0047 736-0046 736-0044 736-0057 736-0065 736-0005 732-1527 732-1528 732-1529 732-1530 732-1531 MLBP902-262XP 732-1532 732-1533 MLBP902-261XP 732-1534 732-1535 MLBP903-033
Robotspiss for Tecan®
Pipettespisser
Spiss av høy kvalitet, spesielt beregnet for molekylærbiologisk arbeid.
  • PP, klar eller sort
  • Spiss med ART®-barriere: eliminerer faren for krysskontaminering mellom prøver. Utformet for bruk med DNA, RNA samt radioaktive og smittefarlige prøver
  • Sertifisert RNase-, DNase og pyrogenfri
  • Steril spiss, gammabestrålt
  • Spiss med større åpning: utformet for viskøse prøver
Bestillingsinformasjon: Fås i forpakninger med 10 eller 50 racks (Econopak).
Bestill nå
SPESIFIKASJONER
Environmentally Preferable Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact
Miljøsertifikat IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603)